Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC2LI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564075 Supplier Invitrogen™ Supplier No. PA564075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PENDIGKLHA HSPMELWKKV YEKLFPPKSI NTLKDIKDPA RDPQYAENEV DEMRIQKDLE LEQYKRSSSK SWKQIEL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 86%.

DYNC2LI1 (dynein, cytoplasmic 2, light intermediate chain 1), also known as LIC3, D2LIC or CGI-60, is a 351 amino acid cytoplasmic protein belonging to the dynein light intermediate chain family. DYNC2LI1 may function as a motor for intraflagellar retrograde transport and in cilia biogenesis. The cytoplasmic dynein complex 2 may be composed of a DYNC2H1 homodimer and a number of DYNC2LI1 light intermediate chains. DYNC2LI1 exists as five alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC2LI1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dync2li1
Gene Accession No. Q8TCX1
Gene Alias 4933404O11Rik; CGI-60; Cytoplasmic dynein 2 light intermediate chain 1; D2LIC; Dync2li1; Dynein 2 light intermediate chain; dynein cytoplasmic 2 light intermediate chain 1; dynein, cytoplasmic 2, light intermediate chain 1; LIC3; mD2LIC; RGD1310286
Gene Symbols Dync2li1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DYNC2LI1. Recombinant protein control fragment (Product #RP-103477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.