Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNC2LI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598717
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598717 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598717 Supplier Invitrogen™ Supplier No. PA598717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYNC2LI1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

DYNC2LI1 (dynein, cytoplasmic 2, light intermediate chain 1), also known as LIC3, D2LIC or CGI-60, is a 351 amino acid cytoplasmic protein belonging to the dynein light intermediate chain family. DYNC2LI1 may function as a motor for intraflagellar retrograde transport and in cilia biogenesis. The cytoplasmic dynein complex 2 may be composed of a DYNC2H1 homodimer and a number of DYNC2LI1 light intermediate chains. DYNC2LI1 exists as five alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNC2LI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dync2li1
Gene Accession No. Q8K0T2, Q8TCX1
Gene Alias 4933404O11Rik; CGI-60; Cytoplasmic dynein 2 light intermediate chain 1; D2LIC; Dync2li1; Dynein 2 light intermediate chain; dynein cytoplasmic 2 light intermediate chain 1; dynein, cytoplasmic 2, light intermediate chain 1; LIC3; mD2LIC; RGD1310286
Gene Symbols Dync2li1
Host Species Rabbit
Immunogen Recombinant Human Cytoplasmic dynein 2 light intermediate chain 1 protein (1-352AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213575, 51626
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.