Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ 3GX™ Dyneema™ Diamond Blended Glove, Latex Coated Palm

Catalog No. 19152923 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Black
Brown
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-152-923 Medium Yellow
19-152-924 Large Brown
19-152-925 X-Large Black
19-152-922 Small Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-152-923 Supplier PIP™ Supplier No. 19D313/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight Dyneema™ blended shell with latex Latex crinkle coating provides superior grip in wet or dry conditions, and offers abrasion resistance.

  • Seamless construction offers increased comfort and breathability
  • Dyneema™ diamond blended shell is lightweight and provides dexterity, tactile sensitivity and cut resistance
  • Latex crinkle coating provides superior grip in wet/dry conditions and offers abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded cuff hems for easy size identification
  • Washable, resistant to chemicals, water and ultraviolet light
  • Compliant with FDA Part 177 - Indirect food activities: Polymers. Subpart C - Substances for use only as components of articles intended for repeated use. Sec. 177.2600

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM F2992-15 ANSI Cut Level A4 (Adopted by ANSI/ISEA 02/16)/ASTM F1790-05 ANSI Cut Level 4
Coating Material Latex
Color White, Blue
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free No
Material Dyneema, Latex
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:3544
For Use With (Application) Electronics, glass cutting, sheet metal handling, parts assembly, construction, general duty applications
Glove Cuff Color Yellow
Glove Cuff Style Knit
Grip Design Crinkle Finish
Model 19-D313/M
Series G-Tek™ 3GX™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.