Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ 3GX™ Dyneema™ Diamond Blended Gloves, Nitrile Coated Palm and Knuckles

Catalog No. 19152918 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Black
Brown
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-152-918 Small Red
19-152-919 Medium Yellow
19-152-920 Large Brown
19-152-921 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-152-918 Supplier PIP™ Supplier No. 19D355/S
This item is not returnable. View return policy

Lightweight Dyneema™ Diamond blended shell provides dexterity, tactile sensitivity and cut resistance. Nitrile coated palm and knuckles are compatible with light oils and provide a secure grip.

  • Seamless construction offers increased comfort and breathability
  • Dyneema™ diamond blended shell is lightweight and provides dexterity, tactile sensitivity and cut resistance
  • Foam nitrile palm and fingertips are compatible with light oils and will provide a secure grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Color coded cuff hems for easy size identification
  • Washable, resistant to chemicals, water and ultraviolet light

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance ASTM F2992-15 ANSI Cut Level A3 (Adopted by ANSI/ISEA 02/16)/ASTM F1790-05 ANSI Cut Level 3
Coating Material Nitrile
Color Salt and Pepper
Cut Resistance Rating ANSI A3
Material Dyneema Diamond, Nitrile
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4443
For Use With (Application) Electronics, glass cutting, sheet metal handling, parts assembly, construction, general duty
Glove Cuff Color Red
Glove Cuff Style Knit
Model 19-D355/S
Series G-Tek™ 3GX™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.