Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Dynein, HC, cytosolic, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABT266MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABT266MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT266MI Supplier MilliporeSigma™ Supplier No. ABT266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytoplasmic dynein 1 heavy chain 1 (UniProt Q14204; also known as Cytoplasmic dynein heavy chain 1, Dynein heavy chain, cytosolic) is encoded by the DYNC1H1 (also known as CMT2O, DHC1, DNCH1, DNCL, DNECL, DYHC, KIAA0325, SMALED1) gene (Gene ID 1778) in human. Cytoplasmic dynein complexes are microtubules-associated retrograde transport motors. Cytoplasmic dynein 1 is the more abundant motor in cells, while cytoplasmic dynein 2 takes part in intraflagellar transport. Cytoplasmic dynein 1 is a multisubunit complex of ∽1.5 MDa composed of a homodimer of heavy chains (encoded by DYNC1H1), two intermediate chains (encoded by DYNC1I1 and DYNC1I2), light-intermediate chains (encoded by DYNC1LI1, DYNC1LI2) and light chains (encoded by DYNLT1, DYNLT3, DYNLRB1, DYNLRB2, DYNLL1, DYNLLl2). The heavy chain homodimer acts as the dynein complex core and binds microtubules to enable ATP-dependent cytoplasmic dynein movement. The other dynein subunits associate as homodimers and play regulatory roles in maintaining the complex stability, modulating its activity, and mediating its interaction with accessory and cargo proteins. The intermediate chain proteins (IC1 and IC2) are involved in cargo binding and specificity, either alone or via interaction with the dynactin complex. IC1 and IC2 share 69% protein identity. They interact with the dynein light chains and the p150 subunit of dynactin at the N-terminus and with the heavy chains through WD40 repeats at the intermediate chain C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynein, HC, cytosolic
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene DYNC1H1, CMT2O, DHC1, DNCH1, DNCL, DNECL, DYHC, KIAA0325, SMALED1
Gene Accession No. Q14204
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence within the second AAA domain of human cytoplasmic dynein heavy chain.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001367
Test Specificity This polyclonal antibody targets a sequence within the second AAA domain of cytoplasmic dynein heavy chain encoded by the DYNC1H1 gene.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.