Learn More
Dynein light chain 2a cytoplasmic Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Dynein light chain 2a cytoplasmic |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | BITH, Bithoraxoid-like protein, DNCL2Acytoplasmic dynein light chain 2A, DNLC2ABLP, Dynein light chain 2A, cytoplasmic, dynein light chain roadblock-type 1, dynein, cytoplasmic, light polypeptide 2A, dynein, light chain, roadblock-type 1, dynein-associated protein HKM23, Dynein-associated protein Km23, roadblock domain containing 1, Roadblock domain-containing protein 1, ROBL/LC7-like 1, ROBLD1Roadblock-1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human Dynein light chain 2a cytoplasmic (NP_054902). Peptide sequence VNTEGIPIKSTMDNPTTTQYASLMHSFILKARSTVRDIDPQNDLTFLRIR |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.