Learn More
Invitrogen™ DYNLL1 Polyclonal Antibody

Description
DYNLL1 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | DYNLL1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | DYNLL1 |
| Gene Accession No. | P63167 |
| Gene Alias | 8 kDa dynein light chain; 8kD LC; 8kDa LC; 8kDLC; CDLC2; cytoplasmic dynein light polypeptide; cytoplasmic dynein light polypeptide 1; DLC1; DLC1DNCLC1; DLC8; DLC8MGC126137; DNCL1; DNCL1MGC126138; Dnclc1; dynein LC8; dynein light chain 1, cytoplasmic; dynein light chain 2; dynein light chain LC8-type 1; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; dynein, light chain, LC8-type 2; Dynll1; dynll2; H LC8; hdlc1; LC8; LC8a; LC8-type 1; MGC126137; MGC126138; mPIN; PIN; PINLC8a; Protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase; similar to cytoplasmic dynein light chain 1; wu:fd56c09; zgc:73406 |
| Gene Symbols | DYNLL1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.