Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNLL1 Recombinant Rabbit Monoclonal Antibody (20A1)
GREENER_CHOICE

Catalog No. FEMA550177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550177 Supplier Invitrogen™ Supplier No. MA550177
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

DYNLL1 Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA

Cytoplasmic dyneins are large enzyme complexes with a molecular mass of about 1, 200 kD. They contain two force-producing heads formed primarily from dynein heavy chains, and stalks linking the heads to a basal domain, which contains a varying number of accessory intermediate chains. The complex is involved in intracellular transport and motility. DYNLL1 is a light chain and exists as part of this complex but also physically interacts with and inhibits the activity of neuronal nitric oxide synthase. Binding of this protein destabilizes the neuronal nitric oxide synthase dimer, a conformation necessary for activity, and it may regulate numerous biologic processes through its effects on nitric oxide synthase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNLL1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 20A1
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DYNLL1
Gene Accession No. P63167
Gene Alias 8 kDa dynein light chain; 8kD LC; 8kDa LC; 8kDLC; CDLC2; cytoplasmic dynein light polypeptide; cytoplasmic dynein light polypeptide 1; DLC1; DLC1DNCLC1; DLC8; DLC8MGC126137; DNCL1; DNCL1MGC126138; Dnclc1; dynein LC8; dynein light chain 1, cytoplasmic; dynein light chain 2; dynein light chain LC8-type 1; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; dynein, light chain, LC8-type 2; Dynll1; dynll2; H LC8; hdlc1; LC8; LC8a; LC8-type 1; MGC126137; MGC126138; mPIN; PIN; PINLC8a; Protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase; similar to cytoplasmic dynein light chain 1; wu:fd56c09; zgc:73406
Gene Symbols DYNLL1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human DYNLL1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8655
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.