Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYNLRB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568174
Description
Predicted to react with Mouse and Rat samples.
The DYNLRB1 gene is a member of the roadblock dynein light chain family. The encoded cytoplasmic protein is capable of binding intermediate chain proteins, interacts with transforming growth factor-beta, and has been implicated in the regulation of actin modulating proteins. Upregulation of this gene has been associated with hepatocellular carcinomas, suggesting that this gene may be involved in tumor progression. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene have been defined on chromosomes 12 and 18.
Specifications
DYNLRB1 | |
Polyclonal | |
Unconjugated | |
Dynlrb1 | |
2010012N15Rik; 2010320M17Rik; 9430076K19Rik; AV124457; BITH; bithoraxoid-like protein; BLP; Dncl2a; DNLC2A; dynein light chain 2A, cytoplasmic; Dynein light chain roadblock-type 1; dynein, cytoplasmic, light chain 2A; dynein, cytoplasmic, light polypeptide 2A; dynein, light chain, roadblock-type 1; dynein-associated protein Km23; dynein-associated protein RKM23; DYNLRB1; HSPC162; hypothetical protein LOC513342; km23; km23-1; Roadblock domain-containing protein 1; ROBL/LC7-like 1; robl/LC7-like protein; ROBLD1; similar to dynein, cytoplasmic, light polypeptide 2A; zgc:56335 | |
Rabbit | |
affinity chromatography | |
RUO | |
170714, 67068, 83658 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62627, P62628, Q9NP97 | |
Dynlrb1 | |
Synthesized peptide derived from human DYNLRB1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction