Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNLRB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103809 Supplier Invitrogen™ Supplier No. PA5103809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DYNLRB2.

Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNLRB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DYNLRB2
Gene Accession No. Q8TF09, Q9DAJ5
Gene Alias 1700009A04Rik; bithoraxoid-like protein; DNCL2B; DNLC2B; Dynein light chain 2B, cytoplasmic; dynein light chain roadblock-type 2; dynein, cytoplasmic, light chain 2B; dynein, cytoplasmic, light polypeptide 2B; dynein, light chain, roadblock-type 2; DYNLRB2; Roadblock domain-containing protein 2; ROBLD2; testicular tissue protein Li 53
Gene Symbols DYNLRB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DYNLRB2(Accession Q8TF09), corresponding to amino acid residues K9-D59.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75465, 83657
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.