Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYNLRB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523763
Description
This antibody is predicted to react with bovine based on sequence homology.
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules.
Specifications
DYNLRB2 | |
Polyclonal | |
Unconjugated | |
DYNLRB2 | |
1700009A04Rik; bithoraxoid-like protein; DNCL2B; DNLC2B; Dynein light chain 2B, cytoplasmic; dynein light chain roadblock-type 2; dynein, cytoplasmic, light chain 2B; dynein, cytoplasmic, light polypeptide 2B; dynein, light chain, roadblock-type 2; DYNLRB2; Roadblock domain-containing protein 2; ROBLD2; testicular tissue protein Li 53 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
75465, 83657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8TF09, Q9DAJ5 | |
DYNLRB2 | |
KLH conjugated synthetic peptide between 49-77 amino acids from the central region of human DYNLRB2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction