Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYNLT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109669 Supplier Invitrogen™ Supplier No. PA5109669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEVGFNAEEA HNIVKECVDG VLGGEDYNHN NINQWTASIV EQSLTHLVKL GKAYKYIVTC AVVQKSAYGF HTASSCFWDT TSDGTCTVRW E.

This gene encodes a member of a subclass of dynein light chains. The encoded protein homodimerizes and forms the light chain component of the cytoplasmic dynein motor protein complex. This protein may be important for binding dynein to specific cargos including the spindle checkpoint protein BUB3. This protein may also function independently of dynein as a transcriptional modulator. Pseudogenes of this gene are found on chromosomes 2 and 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNLT3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DYNLT3
Gene Accession No. P51808, P56387
Gene Alias 2310075M16Rik; AU042796; Dynein light chain Tctex-type 3; dynein, light chain, Tctex-type 3; DYNLT3; Protein 91/23; RP3; T-complex associated-testis-expressed 1-like (Protein 91/23); T-complex-associated testis-expressed 1-like; TCTE1L; TCTE1XL; TCTEX1L
Gene Symbols DYNLT3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 9-99 of human DYNLT3 (NP_006511.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67117, 6990
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.