Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYRK1A Antibody [CoraFluor™ 1], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF5407CL1
Description
DYRK1A Polyclonal antibody specifically detects DYRK1A in Human,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western BlotSpecifications
DYRK1A | |
Polyclonal | |
PBS | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
CoraFluor 1 | |
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A, DYRK1, MNBEC 2.7.110EC 2.7.12 | |
E. coli-derived recombinant human DYRK1A, Arg438-Thr511, Accession # Q13627 | |
0.1 mL | |
Protein Kinase | |
1859 | |
Store at 4°C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction