Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYRK1A Mouse anti-Human, Unconjugated, Clone: 11D9.1, MilliporeSigma™

Catalog No. MABN156425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1564-25 25 μg
MABN1564-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN1564-25 Supplier MilliporeSigma Supplier No. MABN156425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-DYRK1A, clone 11D9.1, Cat. No. MABN1564, is a highly specific mouse monoclonal antibody that targets DYRK1A and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK1A
Applications Western Blot
Classification Monoclonal
Clone 11D9.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Dual specificity tyrosine-phosphorylation-regulated kinase 1A;EC 2.7.12.1;Dual specificity YAK1-related kinase;HP86;Protein kinase minibrain homolog;MNBH;hMNB
Gene Symbols DYRK1A;DYRK;MNB;MNBH
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to the first 95 amino acids from human Dual specificity tyrosine-phosphorylation-regulated kinase 1A (DYRK1A).
Molecular Weight of Antigen ∽86 kDa observed; 85.58 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001387
Test Specificity Clone 11D9.1 specifically detects human Dual specificity tyrosine-phosphorylation-regulated kinase 1A. It targets an epitope within the first 95 amino acids in the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.