Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DYRK1A, Mouse, Unlabeled, Clone: 8D9,

Catalog No. MABN1848 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1848 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1848 Supplier MilliporeSigma™ Supplier No. MABN1848
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dual specificity tyrosine-phosphorylation-regulated kinase 1A (EC 2.7.12.2; UniProt Q63470; also known as Dual specificity YAK1-related kinase, MNBH, Protein kinase minibrain homolog, RP86) is encoded by the Dyrk1a (also known as Dyrk) gene (Gene ID 25255) in rat species. DYRK1A belongs to a family of conserved dual-specificity tyrosine-phosphorylated and regulated kinases (DYRKs) within the CMGC (CDK, MAPK, GSK, and CLK) group of eukaryote kinome. DYRK family members share a conserved kinase domain and an adjacent DYRK-homology domain or DH-box (DDDNXDY), wihile differing in their N- and C-terminal regions. DYRK1A is regulated by autophosphorylation on Ser, Thr and Tyr residues, but catalyzes phosphorylation of a broad-range of substrates only on Ser/Thr residues within the RPX(S/T)P motif with a preference for Pro at the +1 position (X). Known DYRK1A substrates include transcription factors (e.g. FKHR, CREB, Gli1, NFAT), splicing factors (e.g. cycling L2, SF3b/SAP155), glycogen synthase, as well as multiple proteins engaged in endocytosis in neurons, including dynamin and synaptojanin 1. DYRK1A is also implicated in promoting the formation of neurotoxic Aβ peptides by phosphorylating APP and presenilin-1. Human DYRK1A gene is located in the Down syndrome critical region of chromosome 21 and the protein plays an essential role in the development of the central nervous system. DYRK1A haploinsufficiency and mutations are the causes of intellectual disability (ID), microcephaly and dysmorphic features in human, while transgenic mice carrying extra copies of the gene exhibit learning defects and motor abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK1A
Applications Western Blot
Classification Monoclonal
Clone 8D9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Dyrk1a, Dyrk
Gene Accession No. Q63470
Host Species Mouse
Immunogen His-tagged recombinant rat DYRK1A N-terminal fragment.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_036923
Test Specificity The specificity of target band detection was confirmed by antibody blocking with an excess amount of DYRK1A prior to Western blotting (Murakami, N., et al. (2009). Biochemistry. 48(39):9297-9305).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.