Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYRK1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514490
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514490 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514490 Supplier Invitrogen™ Supplier No. PA514490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.

DYRK1A is a dual specificity kinase involved in cell proliferation. The gene for DYRK1A is located in the Down syndrome critical region of chromosome 21, and has been implicated in the pathogenesis of this disease. DYRK1A catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DYRK1A
Gene Accession No. Q13627, Q61214
Gene Alias 2310043O08Rik; D16Ertd272e; D16Ertd493e; dual specificity tyrosine phosphorylation regulated kinase 1A; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; dual specificity tyrosine-phosphorylation-regulated kinase 1A; Dual specificity YAK1-related kinase; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1a; Dyrk; DYRK1; Dyrk1a; ENSMUSG00000074897; Gm10783; hMNB; HP86; mmb; MNB; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; MP86; MRD7; OTTHUMP00000109090; Protein kinase minibrain homolog; PSK47; RP86; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Gene Symbols DYRK1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 107-136 amino acids from the N-terminal region of human DYRK1A.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13548, 1859
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.