Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYRK1B Polyclonal Antibody

DYRK1B Polyclonal Antibody
Supplier: Thermo Scientific PA517236
Description
DYRK1B Polyclonal Antibody for Western Blot, IP

Specifications
DYRK1B | |
Polyclonal | |
Unconjugated | |
DYRK1B | |
AOMS3; dual specificity tyrosine phosphorylation regulated kinase 1B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; dual specificity tyrosine-phosphorylation-regulated kinase 1B; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; DYRK1B; DYRKB; Minibrain-related kinase; Mirk; Mirk protein kinase | |
Rabbit | |
Affinity chromatography | |
RUO | |
13549, 308468, 9149 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
39 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q9Y463, Q9Z188 | |
DYRK1B | |
Synthetic peptide corresponding to residues near the carboxyl terminus of human DYRK1B. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction