Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYRK1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585318
Description
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (92%), Rhesus Monkey (99%), Bovine (99%). Positive Control: mouse testis, rat testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DYRK1B is a member of the DYRK family of protein kinases and is proposed to be involved in nuclear functions. Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. DYRK1B encodes the integrin beta 4 subunit, a receptor for the laminins. Mutations in DYRK1B are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for DYRK1B.
Specifications
DYRK1B | |
Polyclonal | |
Unconjugated | |
DYRK1B | |
AOMS3; dual specificity tyrosine phosphorylation regulated kinase 1B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; dual specificity tyrosine-phosphorylation-regulated kinase 1B; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; DYRK1B; DYRKB; Minibrain-related kinase; Mirk; Mirk protein kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13549, 308468, 9149 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y463, Q9Z188 | |
DYRK1B | |
Recombinant protein encompassing a sequence within the center region of human DYRK1B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction