Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dyrk1B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987161
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7161 Supplier Bioss Supplier No. BS6259R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen Dyrk1B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DYRK1B
Gene Accession No. Q9Y463
Gene Alias AOMS3; dual specificity tyrosine phosphorylation regulated kinase 1B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; Dual specificity tyrosine-phosphorylation-regulated kinase 1B; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B; Dyrk1b; DYRKB; Minibrain-related kinase; MIRK; Mirk protein kinase
Gene Symbols DYRK1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MIRK/Dyrk1B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9149
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.