Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DYRK1B, GST Tag Recombinant Protein

Catalog No. p-7181029
Encompass_Preferred
Click to view available options
Quantity:
10 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

DYRK1B is a member of the DYRK family of protein kinases and is proposed to be involved in nuclear functions. Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. DYRK1B encodes the integrin beta 4 subunit, a receptor for the laminins. Mutations in DYRK1B are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for DYRK1B.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y463
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 9149
Molecular Weight (g/mol) 96.7 kDa
Name Human DYRK1B, GST Tag
Quantity 10 μg
Gene Alias AOMS3; dual specificity tyrosine phosphorylation regulated kinase 1 B; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1 B; dual specificity tyrosine-phosphorylation-regulated kinase 1 B; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1 B; DYRK1B; DYRKB; Minibrain-related kinase; Mirk; Mirk protein kinase
Common Name DYRK1B
KinaseFamily DYRK Kinase Family, DYRKs
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
System Compatibility Z'-LYTE™
Target Entry DYRK1B
Target Gene dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1 B
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.