Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYRK2 Antibody [PerCP], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF5408C
Description
DYRK2 Polyclonal antibody specifically detects DYRK2 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
DYRK2 | |
Polyclonal | |
PBS | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
8445 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
PerCP | |
dual specificity tyrosine-phosphorylation-regulated kinase 2, dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2, EC 2.7.12, EC 2.7.12.1, FLJ21217, FLJ21365 | |
E. coli-derived recombinant human DYRK2, Met74-Glu180, Accession # Q92630 | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction