Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYRK2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982149
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2149 Supplier Bioss Supplier No. BS12332R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DYRK2, or dual specificity tyrosine-phosphorylation-regulated kinase 2, has a role in the regulation of cellular growth and/or development. In response to DNA damage, DYRK2 regulates p53 phosphorylation on Ser-46 to induce apoptosis. DYRK2 has been proposed to inactivate GYS1 by phosphorylation at Ser-641, and potentially a second phosphorylation site, thus regulating glycogen synthesis. DYRK2 also phosphorylates EIF2B5 at Ser-544, enabling its subsequent phosphorylation and inhibition by GSK3, and may play a more general role in the priming of GSK3 substrates.

Specifications

Antigen DYRK2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DYRK2
Gene Accession No. Q92630
Gene Alias 1810038L18Rik; dual specificity tyrosine phosphorylation regulated kinase 2; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 2; Dual specificity tyrosine-phosphorylation-regulated kinase 2; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2; Dyrk2
Gene Symbols DYRK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DYRK2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8445
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.