Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYRK3 Antibody [Biotin], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF5409B
Description
DYRK3 Polyclonal antibody specifically detects DYRK3 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
DYRK3 | |
Polyclonal | |
PBS | |
Goat | |
Antigen Affinity-purified | |
RUO | |
8444 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
dual specificity tyrosine-phosphorylation-regulated kinase 3, dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3, dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5, DYRK5, EC 2.7.12, EC 2.7.12.1, hYAK3-2, protein kinase Dyrk3, REDKRED, Regulatory erythroid kinase | |
E. coli-derived recombinant human DYRK3, Asn52-Glu167, Accession # O43781 | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction