Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYRK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103282 Supplier Invitrogen™ Supplier No. PA5103282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DYRK3.

DYRK3 belongs to the DYRK family of dual-specificity protein kinases that catalyze phosphorylation of serine/threonine and tyrosine residues. The members of this family share structural similarity, but differ in their substrate specificity, suggesting their involvement in different cellular functions. DYRK3 is a negative regulator of EPO-dependent erythropoiesis, may place an upper limit on red cell production during stress erythropoiesis. Inhibits cell death due to cytokine withdrawal in hematopoietic progenitor cells. May act by regulating CREB/CRE signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DYRK3
Gene Accession No. O43781, Q4V8A3, Q922Y0
Gene Alias BC006704; dual specificity tyrosine phosphorylation regulated kinase 3; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual specificity tyrosine-phosphorylation-regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; dyrk3; dyrk3 {ECO:0000312; DYRK5; EMBL:AAH97474.1}; hYAK3-2; KSR; protein kinase Dyrk3; RED; REDK; regulatory erythroid kinase; RP11-343H5.2; RSU2
Gene Symbols DYRK3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DYRK3(Accession O43781), corresponding to amino acid residues S453-L503.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226419, 304775, 8444
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.