Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DYRK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100911 Supplier Invitrogen™ Supplier No. PA5100911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DYRK4.

Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYRK4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dyrk4
Gene Accession No. Q8BI55, Q9NR20
Gene Alias AW049118; dual specificity tyrosine phosphorylation regulated kinase 4; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dual specificity tyrosine-phosphorylation-regulated kinase 4; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 4; dyrk4; Dyrk4a; Dyrk4b; RAD51 associated protein 1; Rad51ap1
Gene Symbols Dyrk4
Host Species Rabbit
Immunogen A synthesized peptide derived from human DYRK4(Accession Q9NR20), corresponding to amino acid residues L479-V520.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101320, 8798
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.