Learn More
Invitrogen™ Dysadherin Monoclonal Antibody (M031)

Description
Clone M031 mouse monoclonal antibody detects a 35-55 kDa* band on SDS-PAGE immunoblots of human A431, A549, MeWo, MDA-MB-231 cell, as well as human breast, lung, and skin tissues. The antibody works for western blot, immunoprecipitation, ELISA, and immunocytochemistry, as well as detects the FXYD5 protein on live cells.
Specifications
Specifications
| Antigen | Dysadherin |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M031 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | FXYD5 |
| Gene Accession No. | Q96DB9 |
| Gene Alias | DYSAD; Dysadherin; EF-8; FXYD domain containing ion transport regulator 5; FXYD domain-containing ion transport regulator 5; FXYD5; HSPC113; Ion channel homolog RIC; IWU1; IWU-1; KCT1; keratinocytes associated transmembrane protein 1; OIT2; oncoprotein induced transcript 2; oncoprotein-induced protein 2; PRO6241; RIC; UNQ2561/PRO6241 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.