Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dysadherin Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA131506
Description
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX22248-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 in variant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Mouse FXYD5 has been termed RIC (Related to Ion Channel). FXYD2, also known as the gamma subunit of the Na,K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. This gene product, FXYD5, is a glycoprotein that functions in the up-regulation of chemokine production, and it is involved in the reduction of cell adhesion via its ability to down-regulate E-cadherin. It also promotes metastasis, and has been linked to a variety of cancers. Alternative splicing results in multiple transcript variants.
Specifications
Dysadherin | |
Polyclonal | |
Unconjugated | |
FXYD5 | |
DYSAD; Dysadherin; EF-8; FXYD domain containing ion transport regulator 5; FXYD domain-containing ion transport regulator 5; FXYD5; HSPC113; Ion channel homolog RIC; IWU1; IWU-1; KCT1; keratinocytes associated transmembrane protein 1; OIT2; oncoprotein induced transcript 2; oncoprotein-induced protein 2; PRO6241; RIC; UNQ2561/PRO6241 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
53827 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96DB9 | |
FXYD5 | |
Peptide with sequence GKCRQLSRLCRNHCR, from the C Terminus. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction