Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysbindin Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. ABN1653MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1653MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1653MI Supplier MilliporeSigma Supplier No. ABN1653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dysbindin is a highly specific rabbit polyclonal antibody that targets Dysbindin-1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dysbindin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols DTNBP1,BLOC1S8,My031
Host Species Rabbit
Immunogen MBP-tagged full length recombinant human Dysbindin.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_115498.2
Test Specificity This polyclonal antibody detects Dysbindin in various regions of human, mouse, and rat brain.
Target Species Mouse, Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.