Learn More
Dysprosium(III) oxide, REacton™, 99.99% (REO)
Description
Dysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts via reaction with acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2340°C |
| Physical Form | Powder |
| Quantity | 500 g |
| Merck Index | 14,3482 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 373 |
| Percent Purity | 99.99% |
| Grade | Reacton™ |
| Sensitivity | Hygroscopic |
| Density | 7.81 g/mL at 27°C |
| Show More |
Safety and Handling
| EINECSNumber | 215-164-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.