Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystonin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18994625UL
Description
Dystonin Polyclonal specifically detects Dystonin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Dystonin | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
230 kDa bullous pemphigoid antigen, BP240, BPAFLJ30627, BPAG1FLJ32235, Bullous pemphigoid antigen, Bullous pemphigoid antigen 1, bullous pemphigoid antigen 1, 230/240kDa, CATX-15, D6S1101, DKFZp564B2416, DMHCATX15, DT, Dystonia musculorum protein, dystonin, FLJ13425, FLJ21489, FLJ46791, Hemidesmosomal plaque protein, KIAA0465, KIAA0728, KIAA1470, MACF2, trabeculin-beta | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.2mg/mL | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Q03001 | |
DST | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KPRCQNVAEDMDIQLCASILNENSDENENINTMILLDKMHSCSSLEKQQRVNVVQLASPSENNLVTEKSNLPEYTTEIAGKSKENLLNHE | |
25 μL | |
Cancer | |
667 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction