Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrobrevin alpha Antibody (OTI1B2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP203422
Description
Dystrobrevin alpha Monoclonal specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Dystrobrevin alpha | |
Monoclonal | |
0.72 mg/mL | |
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
DTNA | |
Full length human recombinant protein of human DTNA (NP_116761) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
1837 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1B2 | |
Unconjugated | |
Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1 | |
Mouse | |
58.7 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction