Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrobrevin beta Antibody (OTI3A12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270598H
Description
Dystrobrevin beta Monoclonal specifically detects Dystrobrevin beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Dystrobrevin beta | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
DTNB | |
Full length recombinant protein of human Dystrobrevin beta (NP_899205) produced in HEK293T cells. | |
0.1 mL | |
1838.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3A12 | |
Western Blot, Immunohistochemistry | |
beta-dystrobrevin, DTN-B, dystrobrevin beta, dystrobrevin, beta, MGC17163, MGC57126 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction