Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystroglycan Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300292MFV500
Description
Dystroglycan Polyclonal antibody specifically detects Dystroglycan in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Dystroglycan | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1 | |
Produced in rabbits immunized with purified, recombinant Human Dystroglycan (Accession#: NP_004384.4; Met1-Val749) | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific | |
1605 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction