Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystroglycan Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$158.00 - $487.50

Specifications

Antigen Dystroglycan
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16901720
SDP
View Documents
Novus Biologicals
NBP16901720UL
20 μL
Each for $158.00
Only null left
Add to Cart
 
NBP169017
SDP
View Documents
Novus Biologicals
NBP169017
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Dystroglycan Polyclonal specifically detects Dystroglycan in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Dystroglycan
Polyclonal
Rabbit
Q62165
1605
Synthetic peptides corresponding to Dag1 (dystroglycan 1) The peptide sequence was selected from the C terminal of Dag1. Peptide sequence PPSPGSSAAPATEVPDRDPEKSSEDDVYLHTVIPAVVVAAILLIAGIIAM.
Primary
Western Blot
Unconjugated
RUO
A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1
DAG1
IgG
97 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.