Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dystrophia myotonica containing WD repeat motif Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10095001IR
Description
dystrophia myotonica containing WD repeat motif Polyclonal antibody specifically detects dystrophia myotonica containing WD repeat motif in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
dystrophia myotonica containing WD repeat motif | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
D19S593E, DM9, DMRN9, DMR-N9, dystrophia myotonica WD repeat-containing protein, dystrophia myotonica, WD repeat containing, dystrophia myotonica-containing WD repeat motif, Dystrophia myotonica-containing WD repeat motif protein, gene59, Protein 59, Protein DMR-N9 | |
The immunogen recognized by this antibody maps to a region between residue 609 and 659 of human dystrophia myotonica, WD repeat containing using the numbering given in entry NP_004934.1 (GeneID 1762). | |
0.1 mL | |
Cell Cycle and Replication | |
1762 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction