Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Dystrophin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS486R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS486R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS486R1 Supplier Epredia™ Supplier No. MS486R1

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, affinity purification and electron microscopy applications.

Dystrophin is a member of the spectrin/α-actinin family of actin-binding, triple helix rod-containing proteins. It is absent or greatly reduced in individuals with the X-linked recessive Duchenne's muscular dystrophy disorder, as well as in mice with the mdx (murine muscular dystrophy) mutation.

Host Species: Mouse

Clone: 1808

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Chicken, Xenopus and Torpedo. Others not known.

Epitope: Not determined

Immunogen: Acetylcholine receptor (AChR)-enriched membranes and peripheral membrane proteins from Torpedo nobiliana electric organ

Molecular Weight: 427 kDa

Positive Control: Skeletal muscle

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Affinity Purification
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Dystrophin Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1808
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Acetylcholine receptor (AChR)-enriched membranes and peripheral membrane proteins from Torpedo nobiliana electric organ
Quantity 1 mL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat, Xenopus species
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.