Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrophin Antibody (4B10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Dystrophin |
---|---|
Clone | 4B10 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Dystrophin | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
A synthesized peptide derived from Human Dystrophin [UniProt P11532] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4B10 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1756 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title