Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Dystrophin Monoclonal Antibody (C4)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Dystrophin |
---|---|
Clone | C4 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541582
|
Invitrogen™
MA541582 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543846
|
Invitrogen™
MA543846 |
20 μL |
Each for $213.50
|
|
|||||
Description
Dystrophin Monoclonal Antibody for Western Blot
Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.Specifications
Dystrophin | |
1 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P11532 | |
1756 | |
Recombinant protein Dystrophin. The antigen corresponds to amino acid range 253-597 of the target protein. | |
Antibody |
C4 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
DMD | |
apodystrophin-3; apodystrophin-I; BMD; CMD3B; dmd; dmd.1; dmd.1.L; DNADMD1; Dp427; Dp71; Duchenne muscular dystrophy (DMD); DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; Dystrophin; dystrophin Dp40; dystrophin Dp71 isoform; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, gene 1 L homeolog; dystrophin, muscular dystrophy; GS1-19O24.1; mdx; MRX85; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592; pke; utrn; utrophin; XELAEV_18012903mg; X-linked muscular dystrophy | |
DMD | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title