Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrophin Monoclonal Antibody (MANDYS8), Invitrogen™

Catalog No. PIMA549307
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549307 Supplier Thermo Scientific Supplier No. MA549307

Mouse Monoclonal Antibody

Add 0.2 mL of PBS buffer will yield a concentration of 500 μg/mL.

Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dystrophin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MANDYS8
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation ascites diluted in 1.2% sodium acetate with 2MG BSA and 0.01MG sodium azide
Gene DMD
Gene Accession No. P11530, P11531, P11532
Gene Alias apodystrophin-3; apodystrophin-I; BMD; CMD3B; dmd; dmd.1; dmd.1.L; DNADMD1; Dp427; Dp71; Duchenne muscular dystrophy (DMD); DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; Dystrophin; dystrophin Dp40; dystrophin Dp71 isoform; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, gene 1 L homeolog; dystrophin, muscular dystrophy; GS1-19O24.1; mdx; MRX85; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592; pke; utrn; utrophin; XELAEV_18012903mg; X-linked muscular dystrophy
Gene Symbols DMD
Host Species Mouse
Immunogen Recombinant human dystrophin fragment.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100355731, 13405, 1756, 24907
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.