Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dystrophin Recombinant Rabbit Monoclonal Antibody (4B10)
GREENER_CHOICE

Catalog No. FEMA549837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549837 Supplier Invitrogen™ Supplier No. MA549837
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Dystrophin Recombinant Monoclonal Antibody for IHC (P), ELISA

Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dystrophin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 4B10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DMD
Gene Accession No. P11532
Gene Alias apodystrophin-3; apodystrophin-I; BMD; CMD3B; dmd; dmd.1; dmd.1.L; DNADMD1; Dp427; Dp71; Duchenne muscular dystrophy (DMD); DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; Dystrophin; dystrophin Dp40; dystrophin Dp71 isoform; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, gene 1 L homeolog; dystrophin, muscular dystrophy; GS1-19O24.1; mdx; MRX85; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592; pke; utrn; utrophin; XELAEV_18012903mg; X-linked muscular dystrophy
Gene Symbols DMD
Host Species Rabbit
Immunogen A synthesized peptide derived from human Dystrophin.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1756
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.