Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DYX1C1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518517
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.
Specifications
DYX1C1 | |
Polyclonal | |
Unconjugated | |
DNAAF4 | |
1700010I24Rik; b2b811Clo; CILD25; DNAAF4; Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; dyslexia susceptibility 1 candidate 1; dyslexia susceptibility 1 candidate 1 homolog; dyslexia susceptibility 1 candidate 1 homolog (human); dyslexia susceptibility 1 candidate gene 1 protein; Dyslexia susceptibility 1 candidate gene 1 protein homolog; DYX1; Dyx1c1; DYXC1; Edem2; Ekn1; FLJ37882; MGC70618; RD | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
161582 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8WXU2 | |
DNAAF4 | |
Synthetic peptide sequence (PLQVSDYSWQQTKT) corresponding to the N-terminus amino acids of DYX1C1 (aa 2-16). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction