Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYX2/KIAA0319 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981731
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1731 Supplier Bioss Supplier No. BS11862R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a transmembrane protein that contains a large extracellular domain with multiple polycystic kidney disease (PKD) domains. The encoded protein may play a role in the development of the cerebral cortex by regulating neuronal migration and cell adhesion. Single nucleotide polymorphisms in this gene are associated with dyslexia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Nov 2011]

Specifications

Antigen DYX2/KIAA0319
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KIAA0319
Gene Accession No. Q5VV43
Gene Alias 4930451E12Rik; D130043K22Rik; DYLX2; dyslexia susceptibility 2; Dyslexia-associated protein KIAA0319; Dyslexia-associated protein KIAA0319 homolog; DYX2; Kiaa0319; mKIAA0319; neuronal migration; NMIG; RIKEN cDNA D130043K22 gene
Gene Symbols KIAA0319
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DYX2/KIAA0319.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9856
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.