Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DZIP1 Monoclonal Antibody (05), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA542965
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542965 100 μL
PIMA543523 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA542965 Supplier Invitrogen™ Supplier No. MA542965
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human DZIP1

May participate in spermatogenesis via its interaction with DAZ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DZIP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Dzip1
Gene Accession No. Q86YF9
Gene Alias 2510025K24Rik; 2810422M04Rik; DAZ interacting protein 1; DAZ interacting protein testis1; DAZ interacting zinc finger protein 1; DAZ-interacting protein 1 homolog; DAZ-interacting protein 1/2; DZIP; Dzip1; DZIP2; DZIPt1; KIAA0996; zinc finger DAZ interacting protein 1; zinc finger protein DZIP1; zinc-finger protein DZIPt1
Gene Symbols Dzip1
Host Species Mouse
Immunogen A synthetic peptide corresponding to the center region of the Human DZIP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22873
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.