Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DZIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563036
Description
Immunogen sequence: SALEYQLSEI QKSNMQIKSN IGTLKDAHEF KEDRSPYPQD FHNVMQLLDS QESKWTARVQ AIHQEHKKEK GRLLSHIEKL RTSMI Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
May participate in spermatogenesis via its interaction with DAZ.
Specifications
DZIP1 | |
Polyclonal | |
Unconjugated | |
Dzip1 | |
2510025K24Rik; 2810422M04Rik; DAZ interacting protein 1; DAZ interacting protein testis1; DAZ interacting zinc finger protein 1; DAZ-interacting protein 1 homolog; DAZ-interacting protein 1/2; DZIP; Dzip1; DZIP2; DZIPt1; KIAA0996; zinc finger DAZ interacting protein 1; zinc finger protein DZIP1; zinc-finger protein DZIPt1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86YF9 | |
Dzip1 | |
Recombinant protein corresponding to Human DZIP1. Recombinant protein control fragment (Product #RP-92296). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction