Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DZIP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532191 Supplier Invitrogen™ Supplier No. PA532191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, mouse kidney. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

E3 Ubiquitin ligase proteins mediate ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Able to specifically bind RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DZIP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene DZIP3
Gene Accession No. Q7TPV2, Q86Y13
Gene Alias 2310047C04Rik; 2A-HUB; 6430549P11Rik; A230104G20; DAZ interacting protein 3, zinc finger; DAZ interacting zinc finger protein 3; DAZ-interacting protein 3; DAZ-interacting protein 3 homolog; DZIP3; E3 ubiquitin-protein ligase DZIP3; hRUL138; human RNA-binding ubiquitin ligase of 138 kDa; Kiaa0675; PPP1R66; protein phosphatase 1, regulatory subunit 66; RGD1563278; RING-type E3 ubiquitin transferase DZIP3; RNA-binding RING-H2 protein-ubiquitin ligase; RNA-binding ubiquitin ligase of 138 kDa; UURF2; UURF2 ubiquitin ligase; zinc finger DAZ interacting protein 3
Gene Symbols DZIP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DZIP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224170, 9666
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.