Learn More
Invitrogen™ DZIP3 Polyclonal Antibody

Description
Immunogen sequence: ESTMKTYVSKL NAETSRALTA EVYFLQCRRD FGLLHLEQTE KECLNQLARV THMAASNLES LQLKAAVDSW NAIVADVRNK IAFLRTQYNE QINKVKQ Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
Specifications
Specifications
| Antigen | DZIP3 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DZIP3 |
| Gene Accession No. | Q86Y13 |
| Gene Alias | 2310047C04Rik; 2A-HUB; 6430549P11Rik; A230104G20; DAZ interacting protein 3, zinc finger; DAZ interacting zinc finger protein 3; DAZ-interacting protein 3; DAZ-interacting protein 3 homolog; DZIP3; E3 ubiquitin-protein ligase DZIP3; hRUL138; human RNA-binding ubiquitin ligase of 138 kDa; Kiaa0675; PPP1R66; protein phosphatase 1, regulatory subunit 66; RGD1563278; RING-type E3 ubiquitin transferase DZIP3; RNA-binding RING-H2 protein-ubiquitin ligase; RNA-binding ubiquitin ligase of 138 kDa; UURF2; UURF2 ubiquitin ligase; zinc finger DAZ interacting protein 3 |
| Gene Symbols | DZIP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.