Learn More
Invitrogen™ DZIP3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total DZIP3.
Specifications
Specifications
| Antigen | DZIP3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | DZIP3 |
| Gene Accession No. | Q86Y13 |
| Gene Alias | 2310047C04Rik; 2A-HUB; 6430549P11Rik; A230104G20; DAZ interacting protein 3, zinc finger; DAZ interacting zinc finger protein 3; DAZ-interacting protein 3; DAZ-interacting protein 3 homolog; DZIP3; E3 ubiquitin-protein ligase DZIP3; hRUL138; human RNA-binding ubiquitin ligase of 138 kDa; Kiaa0675; PPP1R66; protein phosphatase 1, regulatory subunit 66; RGD1563278; RING-type E3 ubiquitin transferase DZIP3; RNA-binding RING-H2 protein-ubiquitin ligase; RNA-binding ubiquitin ligase of 138 kDa; UURF2; UURF2 ubiquitin ligase; zinc finger DAZ interacting protein 3 |
| Gene Symbols | DZIP3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.