Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(E)-1-Pentenylboronic acid pinacol ester, 98%, Thermo Scientific™

Catalog No. AAH2774906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2774906 5 g
AAH2774914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2774906 Supplier Thermo Scientific Chemicals Supplier No. H2774906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as reagents in Suzuki-Miyaura coupling reactions.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep in freezer under -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161395-96-6
Molecular Formula C11H21BO2
Molecular Weight (g/mol) 196.097
MDL Number MFCD05663882
InChI Key IMIHEZMTQOARIK-CMDGGOBGSA-N
Synonym e-4,4,5,5-tetramethyl-2-pent-1-en-1-yl-1,3,2-dioxaborolane, trans-1-penten-1-ylboronic acid pinacol ester, e-1-pentenylboronic acid pinacol ester, trans-1-pentenyl-1-boronic acid pinacol ester, e-penten-1-ylboronic acid pinacol ester, trans-1-pentenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1e-pent-1-en-1-yl-1,3,2-dioxaborolane, e-2-1-pentenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-pent-1-enylboronic acid pinacol ester, 1-pentenyl-1-boronic acid,pinacol ester
PubChem CID 15211560
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-pent-1-enyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CCCC

Specifications

Density 0.884
Boiling Point 110°C to 111°C (28 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.441
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 196.1
Percent Purity 98%
Chemical Name or Material (E)-1-Pentenylboronic acid pinacol ester
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.