Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(E)-2-Octen-1-ol, Thermo Scientific™

Catalog No. AA1570106
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18409-17-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014057
InChI Key AYQPVPFZWIQERS-VOTSOKGWSA-N
Synonym trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol
PubChem CID 5318599
IUPAC Name (E)-oct-2-en-1-ol
SMILES CCCCCC=CCO

Specifications

Density 0.850g/mL
Flash Point 85° (185°F)
Odor Fruit-like
Refractive Index 1.4465
Quantity 5 g
UN Number UN1987
Formula Weight 128.22
Physical Form Liquid
Chemical Name or Material (E)-2-Octen-1-ol
Hazard Category Warning
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.