Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ E 2012
GREENER_CHOICESDP

Catalog No. 555050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-505-0 50 mg
55-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 55-505-0 Supplier Tocris Bioscience™ Supplier No. 5550/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

γ-secretase modulator; Notch-sparing

E 2012 is a γ-secretase modulator. Lowers Aβ40 and Aβ42 levels and increases Aβ37 levels, in vitro and in vivo. Displays no effect on γ-secretase processing of Notch, EphA4 and EphB2.

Specifications

Chemical Name or Material (3E)-1-[(1S)-1-(4-Fluorophenyl)ethyl]-3-[[3-methoxy-4-(4-methyl-1H-imidazol-1-yl)phenyl]methylene]-2-piperidone
CAS 870843-42-8
Quantity 50 mg
Molecular Formula C25H26FN3O2
Formula Weight 419.49
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.